Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044697.2 | NP_001038162.1 | 6 | 819 | ||
NM_001044698.2 | NP_001038163.1 | 6 | 815 | |||
NM_001044700.2 | NP_001038165.1 | 6 | 771 | |||
NM_001294323.1 | NP_001281252.1 | 6 | 778 | |||
NM_178447.3 | NP_848542.1 | 7 | 856 | |||
GenBank mRNA | AF483520.1 | - | 1 | 219 | ||
AF483521.1 | - | 1 | 219 | |||
AK036776.1 | - | 7 | 846 | |||
AK131978.1 | - | 6 | 766 | |||
AK141471.1 | - | 6 | 795 | |||
BC052029.1 | - | 4 | 519 | |||
BC057171.1 | - | 6 | 785 | |||
BC089506.1 | - | 6 | 750 | 30247022 | ||
Y00850.1 | - | 4 | 651 |