Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ikzf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011770.4 | NP_035900.2 | 8 | 1399 | ||
GenBank mRNA | AF044257.1 | - | 8 | 1294 | ||
AK047925.1 | - | 8 | 1261 | |||
BC138606.1 | - | 8 | 1239 | 8861626 | ||
BC138608.1 | - | 8 | 1239 | 8861628 |