Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011794.3 | NP_035924.2 | 5, 6 | 489 | ||
GenBank mRNA | AF125043.1 | - | 5, 6 | 447 | ||
AK008091.1 | - | 4, 5 | 342 | |||
AK008714.1 | - | 5, 6 | 470 | |||
AK093938.1 | - | 5, 6 | 475 | |||
AK150877.1 | - | 5, 6 | 469 | |||
AK151931.1 | - | 5, 6 | 493 | |||
AK152009.1 | - | 5, 6 | 469 | |||
AK154435.1 | - | 5, 6 | 469 | |||
BC011036.1 | - | 5, 6 | 442 | 4160742 | ||
BY744642.1 | - | 5, 6 | 489 |