Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF145285.1 | - | 5 | 553 | ||
AK076117.1 | - | 5 | 603 | |||
AK145697.1 | - | 5 | 602 | |||
AK149427.1 | - | 5 | 581 | |||
AK151732.1 | - | 5 | 637 | |||
AK151965.1 | - | 5 | 570 | |||
AK152792.1 | - | 5 | 570 | |||
AK153221.1 | - | 5 | 593 | |||
AK153893.1 | - | 5 | 541 | |||
AK167186.1 | - | 5 | 605 | |||
AK172256.1 | - | 5 | 570 | |||
AK193944.1 | - | 4 | 353 | |||
AK194613.1 | - | 1 | 46 | |||
AK200302.1 | - | 2 | 65 | |||
AK204630.1 | - | 4 | 353 | |||
AK208498.1 | - | 4 | 353 | |||
BC048176.1 | - | 5 | 590 | 6825491 | ||
BC119806.1 | - | 5 | 471 | 40107463 | ||
BC122883.1 | - | 5 | 471 | 40107462 | ||
CN527272.1 | - | 5 | 677 | |||
U65313.1 | - | 5 | 553 |