Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF145285.1 | - | 12 | 1503 | ||
AK076117.1 | - | 11 | 1454 | |||
AK145697.1 | - | 12 | 1552 | |||
AK149427.1 | - | 12 | 1531 | |||
AK151732.1 | - | 11 | 1489 | |||
AK151965.1 | - | 11 | 1418 | |||
AK152792.1 | - | 11 | 1422 | |||
AK153221.1 | - | 12 | 1543 | |||
AK153893.1 | - | 12 | 1491 | |||
AK167186.1 | - | 11 | 1457 | |||
AK169571.1 | - | 1 | 13 | |||
AK172256.1 | - | 11 | 1422 | |||
AK194974.1 | - | 1 | 28 | |||
AK220349.1 | - | 1 | 151 | |||
BC048176.1 | - | 11 | 1441 | 6825491 | ||
BC119806.1 | - | 11 | 1322 | 40107463 | ||
BC122883.1 | - | 11 | 1322 | 40107462 | ||
U65313.1 | - | 11 | 1404 |