Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267040.1 | NP_001253969.1 | 6, 7 | 979 | ||
NM_004973.3 | NP_004964.2 | 6, 7 | 1479 | |||
XM_005249089.3 | XP_005249146.1 | 5, 6 | 817 | |||
XM_011514579.2 | XP_011512881.1 | 8, 9 | 1668 | |||
XM_011514584.2 | XP_011512886.1 | 4, 5 | 733 | |||
XM_017010833.1 | XP_016866322.1 | 8, 9 | 1623 | |||
XM_017010834.1 | XP_016866323.1 | 5, 6 | 1337 | |||
XM_017010835.1 | XP_016866324.1 | 4, 5 | 712 | |||
GenBank mRNA | AK292861.1 | - | 6, 7 | 1325 | ||
AK299349.1 | - | 5, 6 | 893 | |||
AK302624.1 | - | 5, 6 | 804 | |||
AK303610.1 | - | 6, 7 | 922 | |||
BC046246.1 | - | 6, 7 | 1196 | 4520786 | ||
U57592.1 | - | 6, 7 | 1158 |