Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lynx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011838.4 | NP_035968.1 | 3 | 371 | ||
GenBank mRNA | AF141377.1 | - | 3 | 345 | ||
AK013827.1 | - | 3 | 368 | |||
AK161907.1 | - | 3 | 401 | |||
AK162920.1 | - | 3 | 394 | |||
AK165295.1 | - | 3 | 375 | |||
AK165314.1 | - | 3 | 375 | |||
BC037541.1 | - | 3 | 418 | 5325123 |