Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011846.4 | NP_035976.3 | 9 | 1420 | ||
GenBank mRNA | AB021224.2 | - | 9 | 1426 | ||
AJ010731.1 | - | 9 | 1424 | |||
AK079221.1 | - | 1192 | ||||
AK143521.1 | - | 9 | 1420 | |||
AK179012.1 | - | 1 | 50 | |||
BC051917.1 | - | 9 | 1420 | 5716458 |