Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162947.1 | NP_001156419.1 | 6 | 538 | ||
NM_011848.4 | NP_035978.2 | 7 | 596 | |||
GenBank mRNA | AF093416.1 | - | 6 | 543 | ||
AF099066.1 | - | 7 | 565 | |||
BU058938.1 | - | 7 | 596 |