Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160163.1 | NP_001153635.1 | 4 | 1277 | ||
NM_001160164.1 | NP_001153636.1 | 3 | 1175 | |||
NM_001160165.1 | NP_001153637.1 | 3 | 1303 | |||
NM_015750.3 | NP_056565.1 | 3 | 1143 | |||
GenBank mRNA | AB028023.1 | - | 3 | 1145 | ||
AB048604.1 | - | 3 | 1400 | |||
AF139059.1 | - | 3 | 1253 | |||
AK009828.1 | - | 2 | 1335 | |||
AK145284.1 | - | 3 | 1176 | |||
BC026408.1 | - | 2 | 931 | |||
BC105657.1 | - | 3 | 1145 | 40046586 | ||
BC105658.1 | - | 3 | 1145 | 40046587 | ||
BC105659.1 | - | 3 | 1145 | 40046588 | ||
BC105661.1 | - | 2 | 1405 | 40046584 |