Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013708.4 | NP_038736.1 | 4 | 718 | ||
GenBank mRNA | AF148129.1 | - | 4 | 580 | ||
AF204053.1 | - | 4 | 718 | |||
AK044718.1 | - | 3 | 850 | |||
AK045788.1 | - | 2 | 461 | |||
AK045859.1 | - | 2 | 461 | |||
AK053560.1 | - | 4 | 686 | |||
BC017521.1 | - | 4 | 712 | 4512088 | ||
BC047150.1 | - | 1 | 1405 | |||
BC057958.1 | - | 1 | 1445 |