Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317851.1 | NP_001304780.1 | 11 | 5800 | ||
NM_016173.4 | NP_057257.1 | 11 | 6101 | |||
XM_005265218.3 | XP_005265275.1 | 9 | 5945 | |||
XM_011533806.2 | XP_011532108.1 | 11 | 4870 | |||
XM_011533807.1 | XP_011532109.1 | 11 | 4755 | |||
XM_011533808.1 | XP_011532110.1 | 11 | 4541 | |||
XM_011533809.2 | XP_011532111.1 | 12 | 4467 | |||
XM_011533810.2 | XP_011532112.1 | 12 | 2510 | |||
XM_011533812.2 | XP_011532114.1 | 9 | 4714 | |||
XM_011533813.2 | XP_011532115.1 | 11 | 4908 | |||
XM_011533814.2 | XP_011532116.1 | 11 | 4832 | |||
GenBank mRNA | AF172244.1 | - | 11 | 5701 |