Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011854.2 | NP_035984.2 | 6 | 1971 | ||
GenBank mRNA | AF068835.1 | - | 6 | 1957 | ||
AK010034.1 | - | 6 | 1313 | |||
AK150080.1 | - | 4 | 762 | |||
AK150492.1 | - | 6 | 1302 | |||
AK157649.1 | - | 6 | 1339 | |||
AK160630.1 | - | 5 | 875 |