Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290707.1 | NP_001277636.1 | 19 | 2287 | ||
NM_011866.2 | NP_035996.2 | 18 | 2241 | |||
GenBank mRNA | AF110507.1 | - | 18 | 2112 | ||
AK039249.1 | - | 19 | 2287 | |||
AK147690.1 | - | 17 | 2158 | |||
AK162221.1 | - | 18 | 1896 | |||
AK162804.1 | - | 16 | 1782 | |||
AY360383.1 | - | 18 | 2241 | |||
BC113201.1 | - | 18 | 2107 | 40048238 |