Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011870.5 | NP_036000.1 | 3, 4 | 441 | ||
GenBank mRNA | AB006463.1 | - | 3, 4 | 222 | ||
AF173010.1 | - | 3, 4 | 220 | |||
AK010345.1 | - | 3, 4 | 238 | |||
AK027915.1 | - | 3, 4 | 253 | |||
AK152805.1 | - | 3, 4 | 273 | |||
AK165472.1 | - | 3, 4 | 226 | |||
BC003714.1 | - | 3, 4 | 227 | 3599986 | ||
BC054385.1 | - | 3, 4 | 246 | 6476137 | ||
CF104297.1 | - | 3, 4 | 449 |