Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011871.2 | NP_036001.1 | 7 | 897 | ||
GenBank mRNA | AF083032.1 | - | 7 | 897 | ||
AK012798.1 | - | 7 | 883 | |||
AK155112.1 | - | 7 | 921 | |||
BC011311.1 | - | 7 | 905 | 3985758 |