Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011874.2 | NP_036004.2 | 11 | 1290 | ||
GenBank mRNA | AK050677.1 | - | 11 | 1268 | ||
AK077507.1 | - | 11 | 1253 | |||
AK153991.1 | - | 11 | 1273 | |||
AK160644.1 | - | 11 | 1252 | |||
AK160718.1 | - | 11 | 1244 | |||
AK167041.1 | - | 11 | 1235 | |||
AK167528.1 | - | 11 | 1245 | |||
AK169224.1 | - | 11 | 1253 | |||
AK169244.1 | - | 11 | 1752 | |||
BC012411.1 | - | 3 | 512 | |||
BC012708.1 | - | 11 | 1251 | 4211677 | ||
BC092265.1 | - | 11 | 1234 | 3671250 | ||
BC094063.1 | - | 6 | 608 | |||
BY385516.1 | - | 3 | 283 | |||
L76223.1 | - | 11 | 1241 |