Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112705.2 | NP_001106176.1 | 19 | 2329 | ||
NM_001294329.1 | NP_001281258.1 | 17 | 2189 | |||
NM_001294330.1 | NP_001281259.1 | 17 | 2137 | |||
NM_001294331.1 | NP_001281260.1 | 19 | 2342 | |||
NM_001294334.1 | NP_001281263.1 | 19 | 2329 | |||
GenBank mRNA | AF045252.1 | - | 19 | 2348 | ||
AF045253.1 | - | 18 | 1776 | |||
AF045254.1 | - | 14 | 1490 | |||
AK089925.1 | - | 17 | 1717 | |||
AK161383.1 | - | 1 | 97 | |||
AK165667.1 | - | 19 | 2332 | |||
AK211763.1 | - | 3 | 323 | |||
AX775743.1 | - | 13 | 1354 | |||
BC066198.1 | - | 17 | 1682 | |||
U56909.1 | - | 10 | 1027 |