Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011906.2 | NP_036036.2 | 3 | 484 | ||
GenBank mRNA | AF051098.1 | - | 3 | 291 | ||
AK089317.1 | - | 3 | 355 | |||
AK155116.1 | - | 3 | 428 | |||
AK164913.1 | - | 3 | 241 | |||
AK172045.1 | - | 3 | 415 | |||
BC010244.1 | - | 1 | 40 | |||
BC028977.1 | - | 3 | 398 | 5363085 | ||
BC057296.1 | - | 3 | 417 |