Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vax2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011912.3 | NP_036042.1 | 3 | 851 | ||
GenBank mRNA | AB221556.1 | - | 3 | 822 | ||
AF028715.1 | - | 3 | 825 | |||
BC090635.1 | - | 3 | 848 | 30942578 | ||
BC116389.1 | - | 3 | 843 | 40101824 | ||
BC116390.1 | - | 3 | 843 | 40101825 | ||
Y17792.1 | - | 3 | 851 |