Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011914.2 | NP_036044.1 | 9 | 1313 | ||
GenBank mRNA | AF101435.1 | - | 9 | 1265 | ||
AK019191.1 | - | 9 | 1379 | |||
AK088384.1 | - | 9 | 1329 | |||
AK144173.1 | - | 1328 | ||||
AK146185.1 | - | 9 | 1365 | |||
BC038003.1 | - | 9 | 1314 | 4912719 |