Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011914.2 | NP_036044.1 | 2 | 345 | ||
GenBank mRNA | AF101435.1 | - | 2 | 303 | ||
AK019191.1 | - | 2 | 410 | |||
AK078760.1 | - | 2 | 345 | |||
AK088384.1 | - | 2 | 361 | |||
AK137949.1 | - | 2 | 338 | |||
AK144173.1 | - | 359 | ||||
AK146185.1 | - | 2 | 397 | |||
BC038003.1 | - | 2 | 346 | 4912719 |