Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039185.1 | NP_001034274.1 | 2 | 337 | ||
NM_001039186.1 | NP_001034275.1 | 2 | 337 | |||
NM_001039187.1 | NP_001034276.1 | 2 | 337 | |||
NM_011926.2 | NP_036056.2 | 2 | 337 | |||
GenBank mRNA | AB236328.2 | - | 2 | 246 | ||
AB236329.1 | - | 2 | 246 | |||
AB236330.2 | - | 2 | 246 | |||
AB236331.2 | - | 2 | 246 | |||
AB236332.2 | - | 2 | 246 | |||
AK085408.1 | - | 460 | ||||
AK085749.1 | - | 2 | 296 | |||
BC016891.1 | - | 2 | 289 | |||
M77196.1 | - | 2 | 315 | |||
M96935.1 | - | 2 | 263 | |||
X15351.1 | - | 2 | 349 | |||
X67278.1 | - | 2 | 349 | |||
X67279.1 | - | 2 | 349 | |||
X67282.1 | - | 2 | 349 |