Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007953.2 | NP_031979.2 | 7 | 2159 | ||
GenBank mRNA | AK008536.1 | - | 1 | 489 | ||
AK019222.1 | - | 7 | 2121 | |||
AK156371.1 | - | 7 | 2135 | |||
BC034165.1 | - | 7 | 2102 | |||
BC039774.1 | - | 7 | 2171 | |||
BC052930.1 | - | 1 | 552 | |||
U85259.1 | - | 7 | 2116 |