Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011937.2 | NP_036067.2 | 3 | 305 | ||
GenBank mRNA | AF088903.1 | - | 3 | 263 | ||
AF160355.1 | - | 3 | 251 | |||
AK080610.1 | - | 3 | 254 | |||
AK166069.1 | - | 3 | 255 | |||
AK172879.1 | - | 2 | 168 | |||
AK192632.1 | - | 250 | ||||
BC014800.1 | - | 3 | 236 | 4220980 | ||
BY092839.1 | - | 3 | 280 | |||
CJ048539.1 | - | 3 | 306 |
Gene Symbol: Slc7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK179059.1 | - | 77 |