Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011941.3 | NP_036071.3 | 28 | 4164 | ||
GenBank mRNA | AB029482.1 | - | 29 | 3978 | ||
AK122323.1 | - | 18 | 3114 | |||
BC011271.1 | - | 7 | 1294 | |||
BC070449.1 | - | 28 | 4164 | 30543913 |