Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 6 | 645 | ||
GenBank mRNA | AK008141.1 | - | 6 | 648 | ||
AK011002.1 | - | 6 | 646 | |||
AK053378.1 | - | 6 | 646 | |||
AK150067.1 | - | 6 | 646 | |||
AK159379.1 | - | 6 | 644 | |||
AK166255.1 | - | 6 | 647 | |||
AK166507.1 | - | 6 | 646 | |||
AK167209.1 | - | 6 | 646 | |||
AK167442.1 | - | 6 | 649 | |||
AK168710.1 | - | 6 | 628 | |||
BC007467.1 | - | 6 | 636 | 3497714 | ||
D87115.1 | - | 6 | 658 | |||
X93150.1 | - | 4 | 380 |