Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 12 | 1975 | ||
NM_001284290.1 | NP_001271219.1 | 10 | 1537 | |||
NM_001293104.1 | NP_001280033.1 | 11 | 1790 | |||
NM_001293105.1 | NP_001280034.1 | 10 | 1647 | |||
NR_120531.1 | 11 | 1890 | ||||
XM_005250297.3 | XP_005250354.1 | 11 | 1817 | |||
XM_011516113.1 | XP_011514415.1 | 11 | 1719 | |||
XM_011516114.1 | XP_011514416.1 | 11 | 1647 | |||
XM_017012091.1 | XP_016867580.1 | 10 | 1566 | |||
XM_017012092.1 | XP_016867581.1 | 10 | 1553 | |||
XM_017012093.1 | XP_016867582.1 | 10 | 1494 | |||
GenBank mRNA | AK096764.1 | - | 4 | 2233 | ||
AK223406.1 | - | 12 | 1874 | |||
AK300223.1 | - | 12 | 1764 | |||
AK303819.1 | - | 10 | 1434 | |||
AK312752.1 | - | 12 | 1891 | |||
BC014142.2 | - | 12 | 1845 | 4662011 | ||
BM719712.1 | - | 3 | 228 | |||
CD014092.1 | - | 11 | 1672 | |||
CD014093.1 | - | 10 | 1530 | |||
CD014094.1 | - | 11 | 1772 | |||
M15182.1 | - | 12 | 1870 |