Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011962.3 | NP_036092.1 | 17 | 2510 | ||
GenBank mRNA | AF046783.1 | - | 17 | 1918 | ||
AK013195.1 | - | 17 | 2056 | |||
AK033360.1 | - | 17 | 2055 | |||
AK088948.1 | - | 17 | 1927 | |||
BC043047.1 | - | 17 | 2509 | 5703967 | ||
BC054734.1 | - | 17 | 2510 | 5720686 |