Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011965.2 | NP_036095.1 | 4 | 325 | ||
GenBank mRNA | AF060088.1 | - | 4 | 249 | ||
AJ272019.1 | - | 4 | 278 | |||
AK088673.1 | - | 4 | 292 | |||
AK135326.1 | - | 4 | 263 | |||
AK137054.1 | - | 4 | 267 | |||
AK144784.1 | - | 4 | 280 | |||
AK161611.1 | - | 4 | 267 | |||
AK162277.1 | - | 4 | 272 | |||
AK167689.1 | - | 4 | 263 | |||
BC005762.1 | - | 4 | 262 | 2655483 | ||
BY108158.1 | - | 4 | 325 |