Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016716.4 | NP_057925.1 | 6 | 964 | ||
GenBank mRNA | AF129738.1 | - | 6 | 854 | ||
AK129174.1 | - | 7 | 1352 | |||
BC027304.1 | - | 6 | 989 | 3981170 |