Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172974.2 | NP_766562.1 | 5 | 543 | ||
GenBank mRNA | AF071317.1 | - | 5 | 521 | ||
AK053615.1 | - | 4 | 413 | |||
AK081309.1 | - | 5 | 543 | |||
AK168740.1 | - | 5 | 502 | |||
AK170140.1 | - | 5 | 502 | |||
AK182444.1 | - | 2 | 165 | |||
AK205287.1 | - | 2 | 165 | |||
AK207190.1 | - | 1 | 76 | |||
AK208348.1 | - | 2 | 165 | |||
AK214314.1 | - | 2 | 165 | |||
AK214430.1 | - | 2 | 165 | |||
BC012659.1 | - | 5 | 507 | 4017012 |