Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012010.3 | NP_036140.1 | 2 | 136 | ||
GenBank mRNA | AJ006587.1 | - | 2 | 126 | ||
AK128949.1 | - | 2 | 135 | |||
AK146900.1 | - | 2 | 177 | |||
AK150095.1 | - | 2 | 143 | |||
AK165866.1 | - | 2 | 138 | |||
AK171317.1 | - | 2 | 137 | |||
BC036993.1 | - | 2 | 134 | |||
BC063755.1 | - | 2 | 136 | 6513251 | ||
BC139234.1 | - | 2 | 136 | 8862256 | ||
BC139235.1 | - | 2 | 136 | 8862257 |
Gene Symbol: Eif2s3x-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076900.1 | - | 563 |