Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253808.1 | NP_001240737.1 | 14 | 1667 | ||
NM_001253809.1 | NP_001240738.1 | 14 | 1663 | |||
NM_012025.7 | NP_036155.1 | 15 | 1798 | |||
GenBank mRNA | AB030252.1 | - | 14 | 1647 | ||
AF079974.1 | - | 14 | 1608 | |||
AF212320.1 | - | 14 | 1634 | |||
AF212321.1 | - | 15 | 1765 | |||
AK144608.1 | - | 15 | 1766 | |||
AK144745.1 | - | 14 | 1628 | |||
AK154929.1 | - | 15 | 1786 | |||
AK165897.1 | - | 14 | 1620 | |||
AK166084.1 | - | 15 | 1756 | |||
AK166094.1 | - | 14 | 1625 | |||
AK168020.1 | - | 14 | 1624 | |||
AK168025.1 | - | 14 | 1624 | |||
AK168170.1 | - | 14 | 1624 | |||
AK168679.1 | - | 14 | 1608 | |||
AK181322.1 | - | 2 | 143 | |||
AK207982.1 | - | 2 | 143 | |||
BC010715.1 | - | 14 | 1613 | 3602242 |