Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033988.2 | NP_001029160.1 | 8 | 1065 | ||
NM_001284319.1 | NP_001271248.1 | 8 | 1057 | |||
NM_019744.4 | NP_062718.2 | 8 | 975 | |||
GenBank mRNA | AF159461.1 | - | 8 | 972 | ||
AK032951.1 | - | 6 | 731 | |||
AK129020.1 | - | 8 | 1023 | |||
AK143965.1 | - | 8 | 969 | |||
AK145070.1 | - | 8 | 1026 | |||
AK145945.1 | - | 8 | 1018 | |||
AK145954.1 | - | 8 | 1022 | |||
AK146384.1 | - | 8 | 976 | |||
AK146498.1 | - | 8 | 1015 | |||
AK146598.1 | - | 8 | 999 | |||
AK146646.1 | - | 8 | 970 | |||
AK146649.1 | - | 8 | 972 | |||
AK147036.1 | - | 8 | 973 | |||
AK147125.1 | - | 8 | 1017 | |||
AK156999.1 | - | 8 | 1057 | |||
BC031528.1 | - | 8 | 1016 | 4008986 | ||
BC085086.1 | - | 8 | 961 | 6335192 |