Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164495.1 | NP_001157967.1 | 4 | 654 | ||
NM_013722.3 | NP_038750.2 | 4 | 654 | |||
GenBank mRNA | AF192754.1 | - | 4 | 599 | ||
AF498252.1 | - | 3 | 401 | |||
AF498253.1 | - | 3 | 401 | |||
AK043854.1 | - | 4 | 655 | |||
AK048643.1 | - | 4 | 692 | |||
AK080557.1 | - | 4 | 615 | |||
AK082445.1 | - | 4 | 692 | |||
AK084032.1 | - | 4 | 640 | |||
AK086955.1 | - | 601 | ||||
AK087342.1 | - | 5 | 730 | |||
BC107397.2 | - | 3 | 520 | 40058225 |