Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013724.2 | NP_038752.2 | 25 | 4261 | ||
GenBank mRNA | AB020741.1 | - | 25 | 4186 | ||
AB035267.1 | - | 25 | 3939 | |||
BC139029.1 | - | 25 | 4028 | 8862051 | ||
BC139031.1 | - | 25 | 4028 | 8862053 |