Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013724.2 | NP_038752.2 | 13 | 1920 | ||
GenBank mRNA | AB020741.1 | - | 13 | 1845 | ||
AB035267.1 | - | 13 | 1598 | |||
BC068311.1 | - | 13 | 1895 | |||
BC139029.1 | - | 13 | 1687 | 8862051 | ||
BC139031.1 | - | 13 | 1687 | 8862053 |