Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013737.5 | NP_038765.2 | 8 | 1076 | ||
GenBank mRNA | AK005158.1 | - | 8 | 813 | ||
AK051454.1 | - | 8 | 1077 | |||
AK155678.1 | - | 9 | 927 | |||
BC010726.1 | - | 8 | 832 | 3707891 | ||
U34277.1 | - | 8 | 822 |