Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199004.1 | NP_001185933.1 | 11 | 2269 | ||
NM_013747.4 | NP_038775.1 | 11 | 2111 | |||
GenBank mRNA | AB016784.1 | - | 11 | 2270 | ||
AF026274.1 | - | 11 | 2101 | |||
AK138455.1 | - | 11 | 2270 | |||
AK168765.1 | - | 11 | 2115 | |||
BC016883.1 | - | 11 | 2839 | 4235952 | ||
BC086782.1 | - | 11 | 2111 | 6813878 |