Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013753.2 | NP_038781.2 | 19 | 2611 | ||
GenBank mRNA | AK088577.1 | - | 19 | 2447 | ||
AK146489.1 | - | 19 | 2556 | |||
BC137695.1 | - | 18 | 2375 | 8860714 | ||
BC144846.1 | - | 18 | 2384 | 9053394 | ||
BC144847.1 | - | 18 | 2381 | 9053395 | ||
X99384.1 | - | 18 | 2396 |