Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013770.2 | NP_038798.2 | 4 | 511 | ||
GenBank mRNA | AF188712.1 | - | 4 | 482 | ||
AK008038.1 | - | 4 | 481 | |||
AK050282.1 | - | 4 | 465 | |||
AK083831.1 | - | 4 | 489 | |||
AK151645.1 | - | 4 | 511 | |||
BC003222.1 | - | 4 | 429 | 3482316 |