Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013782.4 | NP_038810.2 | 10 | 1145 | ||
GenBank mRNA | AF099053.1 | - | 10 | 1029 | ||
AK036368.1 | - | 10 | 1146 | |||
AK048026.1 | - | 10 | 1128 | |||
AK081275.1 | - | 10 | 1112 | |||
AK137415.1 | - | 1106 | ||||
BC009013.1 | - | 8 | 812 | |||
BC053517.1 | - | 10 | 1082 | 6315175 |