Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175094.5 | NP_780303.1 | 8 | 1054 | ||
GenBank mRNA | AK047670.1 | - | 8 | 1052 | ||
AK050433.1 | - | 8 | 1055 | |||
AK083170.1 | - | 8 | 1018 | |||
AK166257.1 | - | 8 | 1017 | |||
BC061231.1 | - | 8 | 1015 | 6532760 |