Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013791.2 | NP_038819.1 | 1, 2 | 132 | ||
GenBank mRNA | AK037967.1 | - | 1, 2 | 133 | ||
AK077541.1 | - | 1, 2 | 128 | |||
AK155457.1 | - | 1, 2 | 113 | |||
AK164822.1 | - | 1, 2 | 128 | |||
AK165239.1 | - | 1, 2 | 132 | |||
AK171980.1 | - | 1, 2 | 116 | |||
BC013703.1 | - | 1, 2 | 116 | 3668925 | ||
CD550341.1 | - | 1, 2 | 225 | |||
U72194.1 | - | 1, 2 | 115 |