Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018795.2 | NP_061265.2 | 22 | 2796 | ||
GenBank mRNA | AB028737.1 | - | 22 | 2802 | ||
BC040400.1 | - | 22 | 2804 | |||
BC049980.1 | - | 6 | 739 | |||
EF109740.1 | - | 22 | 2796 | |||
EF109741.1 | - | 22 | 2796 |