Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138584.2 | NP_613050.1 | 3, 4 | 465 | ||
GenBank mRNA | AK009332.1 | - | 3, 4 | 457 | ||
AK011985.1 | - | 3, 4 | 432 | |||
AK090017.1 | - | 3, 4 | 453 | |||
AK149930.1 | - | 3, 4 | 439 | |||
AK150215.1 | - | 3, 4 | 466 | |||
AK150581.1 | - | 3, 4 | 443 | |||
AK153284.1 | - | 3, 4 | 427 | |||
AK170666.1 | - | 3, 4 | 458 | |||
BC099434.1 | - | 2, 3 | 230 | 30714184 | ||
BC132377.1 | - | 3, 4 | 442 | 40130654 | ||
BC132403.1 | - | 3, 4 | 442 | 40130680 |