Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003799.1 | - | 3 | 1310 | ||
AK028145.1 | - | 3 | 1312 | |||
AK029529.1 | - | 3 | 1309 | |||
AK160993.1 | - | 3 | 1310 | |||
AK169760.1 | - | 3 | 1310 | |||
BC043057.1 | - | 3 | 1294 | 6419597 | ||
BC046231.1 | - | 3 | 1211 | 6813388 | ||
BC054728.1 | - | 3 | 1282 | 6419723 | ||
BC056986.1 | - | 3 | 1243 | 6816649 |