Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138586.3 | NP_613052.1 | 6 | 976 | ||
GenBank mRNA | AK006475.1 | - | 6 | 929 | ||
AK007372.1 | - | 6 | 965 | |||
AK155228.1 | - | 6 | 928 | |||
BC034358.1 | - | 6 | 922 | 2651418 | ||
BC092393.1 | - | 2 | 5013 |