Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138596.2 | NP_613062.1 | 3, 4 | 363 | ||
GenBank mRNA | AK003862.1 | - | 3, 4 | 357 | ||
AK013992.1 | - | 3, 4 | 367 | |||
AK156498.1 | - | 3, 4 | 372 | |||
AK215905.1 | - | 3, 4 | 211 | |||
BC048638.1 | - | 3, 4 | 356 | 6771841 | ||
BC056438.1 | - | 3, 4 | 352 | 6818638 |